Learn More
ZM 449829, Tocris Bioscience™
Potent, selective JAK3 inhibitor
Brand:  Tocris (Bio-Techne) 136610
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13469233
  • £127.89 / 10mg
Estimated Shipment date
from Supplier 25-03-2020
Description
Description
Potent, selective inhibitor of Janus tyrosine kinase 3 (JAK3) which binds competitively to the JAK3 ATP site. pIC50 values are 6.8, 5.0, 4.7, and < 5.0 for JAK3, EGFR, JAK1 and CDK4 respectively. Inhibits STAT-5 phosphorylation and T cell proliferation.
Specifications
Chemical Identifiers
4452-06-6
182.22
1-naphthalen-2-yl prop-2-en-1-one, 1-2-naphthyl prop-2-en-1-one, jak3 inhibitor v, 2-naphthylvinyl ketone, 2'-acrylonaphthone, 2-acryloylnaphthalene, thr1 inhibitor, 1, 2-naphthyl vinyl ketone, biomolki_000064, biomolki2_000005
1-naphthalen-2-ylprop-2-en-1-one
C13H10O
FOTCGZPFSUWZBN-UHFFFAOYSA-N
3799
C=CC(=O)C1=CC2=CC=CC=C2C=C1
Specifications
10mg
4452-06-6
1-naphthalen-2-yl prop-2-en-1-one, 1-2-naphthyl prop-2-en-1-one, jak3 inhibitor v, 2-naphthylvinyl ketone, 2'-acrylonaphthone, 2-acryloylnaphthalene, thr1 inhibitor, 1, 2-naphthyl vinyl ketone, biomolki_000064, biomolki2_000005
C=CC(=O)C1=CC2=CC=CC=C2C=C1
182.22
182.22
ZM 449829
C13H10O
FOTCGZPFSUWZBN-UHFFFAOYSA-N
1-naphthalen-2-ylprop-2-en-1-one
3799
>98%