missing translation for 'onlineSavingsMsg'
Learn More

ZM 241385, Tocris Bioscience™

Potent, highly selective A2A antagonist

Brand:  Tocris (Bio-Techne) 103650

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13488213

  • £557.72 / 50mg


Estimated Shipment
04-10-2018
Add to basket

Description

Description

Potent and highly selective A2A adenosine antagonist, with a pA2 of 9.02 for A2A receptors in guinea pig cardiac vasculature and selectivities of 1000, 91 and 500,000 over A1, A2B and A3 sites respectively. Active in vivo.
Specifications

Chemical Identifiers

139180-30-6
337.343
4-(2-[7-Amino-2-(2-furyl)[1,2,4]triazolo[2,3-a][1,3,5]triazin-5-ylamino]ethyl)phenol
4-[2-[[7-amino-2-(furan-2-yl)-[1,2,4]triazolo[1,5-a][1,3,5]triazin-5-yl]amino]ethyl]phenol
C16H15N7O2
PWTBZOIUWZOPFT-UHFFFAOYSA-N
176407
C1=COC(=C1)C2=NN3C(=NC(=NC3=N2)NCCC4=CC=C(C=C4)O)N

Specifications

ZM 241385
>99%
50mg
PWTBZOIUWZOPFT-UHFFFAOYSA-N
4-[2-[[7-amino-2-(furan-2-yl)-[1,2,4]triazolo[1,5-a][1,3,5]triazin-5-yl]amino]ethyl]phenol
176407
139180-30-6
C16H15N7O2
4-(2-[7-Amino-2-(2-furyl)[1,2,4]triazolo[2,3-a][1,3,5]triazin-5-ylamino]ethyl)phenol
C1=COC(=C1)C2=NN3C(=NC(=NC3=N2)NCCC4=CC=C(C=C4)O)N
337.343
337.34
Videos