Learn More
Tocris Bioscience™ ZM 226600
Brand:  Tocris Bioscience™ 088210
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13468103
  • £119.07 / 10mg
Estimated Shipment date
from Supplier 03-04-2020
Description
Description
Potent Kir6 (KATP) channel opener (EC50 = 0.5 μM), devoid of antiandrogen properties.
Specifications
Specifications
N-(4-Phenylsulfonylphenyl)-3,3,3-trifluoro-2-hydroxy-2-methylpropanamide
C16H14F3NO4S
10mg
147695-92-9
Observed MW: 373.35
Soluble to 100mM in DMSO