missing translation for 'onlineSavingsMsg'
Learn More

ZK 93423 hydrochloride, Tocris Bioscience™

Potent benzodiazepine agonist

Brand:  Tocris (Bio-Techne) 199450

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13460364

  • £600.29 / 50mg


Estimated Shipment
07-03-2019
Add to basket

Description

Description

Potent benzodiazepine receptor agonist (IC50 = 1 nM). Non-selective between α1-, α2-, α3- and α5-subunit containing GABAA receptors (Ki values are 4.1, 4.2, 6 and 4.5 nM for inhibition of [3H]Ro15-1788 binding to human recombinant α1β3γ2, α2β3γ2, α3β3γ2 and α5β3γ2 receptors respectively). Anxiolytic following systemic administration in vivo.
Specifications

Chemical Identifiers

1216574-52-5
426.897
zk 93423 hydrochloride, 4-methoxymethyl-6-phenylmethoxy-9h-pyrido 3,4-b indole-3-carboxylic acid ethyl ester hydrochloride, 4-methoxymethyl-6-phenylmethoxy-9h-pyrido 3,4-b indole-3-carboxylic acid ethyl ester, 9h-pyrido 3,4-b indole-3-carboxylicacid, 4-methoxymethyl-6-phenylmethoxy-, ethyl ester
ethyl 4-(methoxymethyl)-6-phenylmethoxy-9H-pyrido[3,4-b]indole-3-carboxylate;hydrochloride
C23H23ClN2O4
KHMDBPAODAAMBQ-UHFFFAOYSA-N
56972166
CCOC(=O)C1=NC=C2C(=C1COC)C3=C(N2)C=CC(=C3)OCC4=CC=CC=C4.Cl

Specifications

ZK 93423 hydrochloride
1216574-52-5
zk 93423 hydrochloride, 4-methoxymethyl-6-phenylmethoxy-9h-pyrido 3,4-b indole-3-carboxylic acid ethyl ester hydrochloride, 4-methoxymethyl-6-phenylmethoxy-9h-pyrido 3,4-b indole-3-carboxylic acid ethyl ester, 9h-pyrido 3,4-b indole-3-carboxylicacid, 4-methoxymethyl-6-phenylmethoxy-, ethyl ester
CCOC(=O)C1=NC=C2C(=C1COC)C3=C(N2)C=CC(=C3)OCC4=CC=CC=C4.Cl
426.897
426.9
50mg
C23H23ClN2O4
KHMDBPAODAAMBQ-UHFFFAOYSA-N
ethyl 4-(methoxymethyl)-6-phenylmethoxy-9H-pyrido[3,4-b]indole-3-carboxylate;hydrochloride
56972166
>98%
Videos