missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZK 756326, Tocris Bioscience™
Selective, non-peptide CCR8 agonist
Brand: Tocris (Bio-Techne) 256550
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity | 50mg |
---|
Description
Selective, non-peptide CCR8 chemokine receptor agonist (IC50 values are 1.8 and 2.6 μM for human and mouse receptors respectively). Displays no activity at CCR4, CXCR3, CXCR4 and CCR5 and shows > 28-fold selectivity over 26 other GPCRs (less selective at α2A and 5-HT receptors). Induces chemotaxis and inhibits Env-mediated (HIV) cell-cell fusion.Chemical Identifiers
874911-96-3 | |
429.382 | |
zk756326 dihydrochloride, acmc-20p1e4, 2-2-4-3-phenoxyphenyl methyl piperazin-1-yl ethoxy ethanol dihydrochloride, 2-2-4-3-phenoxyphenyl methyl-1-piperazinyl ethoxy ethanol dihydrochloride | |
2-[2-[4-[(3-phenoxyphenyl)methyl]piperazin-1-yl]ethoxy]ethanol;dihydrochloride |
C21H30Cl2N2O3 | |
MPACCEKWFGWZHS-UHFFFAOYSA-N | |
56972201 | |
C1CN(CCN1CCOCCO)CC2=CC(=CC=C2)OC3=CC=CC=C3.Cl.Cl |
Specifications
ZK 756326 | |
874911-96-3 | |
zk756326 dihydrochloride, acmc-20p1e4, 2-2-4-3-phenoxyphenyl methyl piperazin-1-yl ethoxy ethanol dihydrochloride, 2-2-4-3-phenoxyphenyl methyl-1-piperazinyl ethoxy ethanol dihydrochloride | |
C1CN(CCN1CCOCCO)CC2=CC(=CC=C2)OC3=CC=CC=C3.Cl.Cl | |
429.382 | |
429.38 |
50mg | |
C21H30Cl2N2O3 | |
MPACCEKWFGWZHS-UHFFFAOYSA-N | |
2-[2-[4-[(3-phenoxyphenyl)methyl]piperazin-1-yl]ethoxy]ethanol;dihydrochloride | |
56972201 | |
>99% |