missing translation for 'onlineSavingsMsg'
Learn More

ZK 756326, Tocris Bioscience™

Selective, non-peptide CCR8 agonist

Brand:  Tocris (Bio-Techne) 256510

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13441484

  • £127.89 / 10mg

Estimated Shipment date
from Supplier 25-10-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Selective, non-peptide CCR8 chemokine receptor agonist (IC50 values are 1.8 and 2.6 μM for human and mouse receptors respectively). Displays no activity at CCR4, CXCR3, CXCR4 and CCR5 and shows > 28-fold selectivity over 26 other GPCRs (less selective at α2A and 5-HT receptors). Induces chemotaxis and inhibits Env-mediated (HIV) cell-cell fusion.
Specifications

Chemical Identifiers

874911-96-3
429.382
zk756326 dihydrochloride, acmc-20p1e4, 2-2-4-3-phenoxyphenyl methyl piperazin-1-yl ethoxy ethanol dihydrochloride, 2-2-4-3-phenoxyphenyl methyl-1-piperazinyl ethoxy ethanol dihydrochloride
2-[2-[4-[(3-phenoxyphenyl)methyl]piperazin-1-yl]ethoxy]ethanol;dihydrochloride
C21H30Cl2N2O3
MPACCEKWFGWZHS-UHFFFAOYSA-N
56972201
C1CN(CCN1CCOCCO)CC2=CC(=CC=C2)OC3=CC=CC=C3.Cl.Cl

Specifications

10mg
874911-96-3
zk756326 dihydrochloride, acmc-20p1e4, 2-2-4-3-phenoxyphenyl methyl piperazin-1-yl ethoxy ethanol dihydrochloride, 2-2-4-3-phenoxyphenyl methyl-1-piperazinyl ethoxy ethanol dihydrochloride
C1CN(CCN1CCOCCO)CC2=CC(=CC=C2)OC3=CC=CC=C3.Cl.Cl
429.382
429.38
ZK 756326
C21H30Cl2N2O3
MPACCEKWFGWZHS-UHFFFAOYSA-N
2-[2-[4-[(3-phenoxyphenyl)methyl]piperazin-1-yl]ethoxy]ethanol;dihydrochloride
56972201
>99%
Videos
Documents