missing translation for 'onlineSavingsMsg'
Learn More

ZK 200775, Tocris Bioscience™

Competitive AMPA/kainate antagonist

Brand:  Tocris (Bio-Techne) 234550

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13461074

  • £684.77 / 50mg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Competitive AMPA/kainate antagonist. In rat cortical membranes, displays high affinity for [3H]-AMPA (Ki = 120 nM) and [3H]-CNQX (Ki = 32 nM) binding sites and low affinity for kainate and NMDA channel-associated binding sites (IC50 values range from 2.5 to 11 μM). Inhibits currents induced by AMPA, kainate and NMDA with IC50 values of 21 nM, 27 nM, and > 1 μM respectively. Displays anxiolytic, anticonvulsant and muscle relaxant activity in vivo.
Specifications

Chemical Identifiers

161605-73-8
409.258
fanapanel, mpqx, fanapanel inn, unii-e3ap71em0o, e3ap71em0o, 7-morpholin-4-yl-2,3-dioxo-6-trifluoromethyl-4h-quinoxalin-1-yl methylphosphonic acid, dsstox_cid_26093, dsstox_rid_81332, dsstox_gsid_46093, 3,4-dihydro-7-morpholino-2,3-dioxo-6-trifluoromethyl-1 2h-quinoxalinyl methyl phosphonic acid
[7-morpholin-4-yl-2,3-dioxo-6-(trifluoromethyl)-4H-quinoxalin-1-yl]methylphosphonic acid
C14H15F3N3O6P
WZMQMKNCWDCCMT-UHFFFAOYSA-N
208953
C1COCCN1C2=CC3=C(C=C2C(F)(F)F)NC(=O)C(=O)N3CP(=O)(O)O

Specifications

ZK 200775
161605-73-8
fanapanel, mpqx, fanapanel inn, unii-e3ap71em0o, e3ap71em0o, 7-morpholin-4-yl-2,3-dioxo-6-trifluoromethyl-4h-quinoxalin-1-yl methylphosphonic acid, dsstox_cid_26093, dsstox_rid_81332, dsstox_gsid_46093, 3,4-dihydro-7-morpholino-2,3-dioxo-6-trifluoromethyl-1 2h-quinoxalinyl methyl phosphonic acid
C1COCCN1C2=CC3=C(C=C2C(F)(F)F)NC(=O)C(=O)N3CP(=O)(O)O
409.258
409.25
50mg
C14H15F3N3O6P
WZMQMKNCWDCCMT-UHFFFAOYSA-N
[7-morpholin-4-yl-2,3-dioxo-6-(trifluoromethyl)-4H-quinoxalin-1-yl]methylphosphonic acid
208953
>99%
Videos