Learn More
Tocris Bioscience™ ZJ 43
Brand:  Tocris Bioscience™ 267510
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13461704
  • £249.49 / 10mg
Estimated Shipment
07-03-2019
Add to basket
Description
Description
Potent inhibitor of glutamate carboxypeptidase II and III (GCP II and III/NAAG peptidase/NAALADase) (Ki values are 0.8 and 23nM respectively) that inhibits the hydrolysis of NAAG (IC50 = 2.4nM) . Does not directly interact with NMDA or metabotropic glutamate receptors. Reduces neuronal degeneration in a rat model of? traumatic brain injury (TBI) and reduces locomotor activity in the PCP-model of schizophrenia.
Specifications
Specifications
N-[[[(1S)-1-Carboxy-3-methylbutyl]amino]carbonyl]-L-glutamic acid
C12H20N2O7
10mg
723331-20-2
Observed MW: 304.3
Soluble to 100mM in DMSO