missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ ZJ-43

Brand:  Alfa Aesar™ J64557.MA

Code : NEW

Additional Details : CAS Number : 723331-20-2 Weight : 0.00010kg

 View more versions of this product

Product Code. 15436459

  • £250.69 / 10mg


Estimated Shipment date
from Supplier 11-06-2018
Add to basket

Description

Description

  • Strong inhibitor of N- acetylaspartylglutamate peptidases, glutamate carboxypeptidase II and glutamate carboxypeptidase III
Specifications

Specifications

10mg
−30° to −10°C
Strong inhibitor of N- acetylaspartylglutamate peptidases, glutamate carboxypeptidase II and glutamate carboxypeptidase III.
C12H20N2O7
(S)-2-(3-((S)-1-carboxy-3-methylbutyl)ureido)pentanedioic acid
White
Solid
723331-20-2
304.30g/mol
Signal Transduction Reagent
Videos
SDS