missing translation for 'onlineSavingsMsg'
Learn More

Zirconium titanium oxide, 99.5% (metals basis excluding Hf), Hf <75ppm, Thermo Scientific Chemicals

Catalog Number p-7050799
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
12036-70-3
MFCD00210648
zirconium titanium oxide, tetrahydrate titanium zirconium
This item is not returnable. View return policy
12036-70-3
MFCD00210648
zirconium titanium oxide, tetrahydrate titanium zirconium

Suitable for glass, optic and ceramic applications, advanced electronics and in light weight structural components in aerospace and electrochemical applications such as fuel cells in which they exhibit ionic conductivity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Powder
Quantity 25 g
Solubility Information Insoluble in water.
Formula Weight 203.11
Percent Purity 99.5%
Odor Odorless
Assay (metals basis excluding Hf), Hf <75ppm
Chemical Name or Material Zirconium titanium oxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'