missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Zirconium plate, 6.4mm (0.25 in.) thick, 50x100mm (1.96x3.9in), 99.2% (metals basis excluding Hf)

Brand:  Alfa Aesar™ 044260.KJ

Code : NEW

Additional Details : CAS Number : 7440-67-7 Weight : 0.01000kg

Quantity 5 piece

 View more versions of this product

Product Code. 15474905

  • £369.52 / Pack of 5


Estimated Shipment date
from Supplier 07-06-2018
Add to basket

Specifications

Chemical Identifiers

Zirconium plate
7440-67-7
6.4mm (0.25 in.) thick, 50x100mm (1.96x3.9 in.), Temper: annealed
MFCD00011303

Specifications

Zirconium plate
5 piece
7440-67-7
99.2%
MFCD00011303
≈208g/50x100mm
6.4mm (0.25 in.) thick, 50x100mm (1.96x3.9 in.), Temper: annealed
Plate
(metals basis excluding Hf)
Odorless
Videos
SDS