missing translation for 'onlineSavingsMsg'
Learn More

Zirconium(IV) oxide, Puratronic™, 99.978% (metals basis), Thermo Scientific Chemicals

Catalog Number 11306168
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
1314-23-4
O2Zr
123.22
MFCD00011310 MFCD01868884
MCMNRKCIXSYSNV-UHFFFAOYSA-N
zirconia, zirconium oxide, zirconium oxide zro2, rhuligel, zirconium iv oxide, zirconium white, zirconic anhydride, pigment white 12, zirox zt 35, pcs filler
62395
dioxozirconium
O=[Zr]=O
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
1314-23-4
O2Zr
123.22
MFCD00011310 MFCD01868884
MCMNRKCIXSYSNV-UHFFFAOYSA-N
zirconia, zirconium oxide, zirconium oxide zro2, rhuligel, zirconium iv oxide, zirconium white, zirconic anhydride, pigment white 12, zirox zt 35, pcs filler
62395
dioxozirconium
O=[Zr]=O

Zirconium(IV) oxide is used as an additive in the production of piezoelectric crystals, in high-frequency induction coils and in ceramic glazes. It finds application as a solid electrolyte in electrochromic devices and precursor to the electroceramic lead zirconate titanate (PZT), as an insulator in transistors, as a thermal barrier coating in jet and diesel engines and as a diamond simulant in jewelry. It is used in oxygen sensors, fuel cell membranes and in orthopaedic implants of hip joints. It is coated on the silica gel surface and found to be used in the extraction of chromium from aqueous solutions at a pH of about 3.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point ∼5000°C
Melting Point ∼2700°C
Color White
Physical Form Powder
Quantity 25 g
Linear Formula ZrO2
Merck Index 14,10180
Solubility Information Soluble in alcohols,hydrofluoric and hot sulfuric acid. Insoluble in water.
Formula Weight 123.22
Percent Purity 99.978%
Grade Puratronic™
Odor Odorless
Density 5.89 g/mL
Assay (metals basis)
Chemical Name or Material Zirconium(IV) oxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'