Learn More
Zirconium(IV) isopropoxide isopropanol complex, Thermo Scientific Chemicals
Description
Zirconium(IV) isopropoxide isopropanol is a suitable precursor for the synthesis of olefin-functionalized zirconium containing monomers. It is used in the manufacture of hybrid zirconium containing polymers. It is also used as a source of zirconia, a well known material having high mechanical strength, chemical durability and alkali resistance. It is also used for the chemical protection of metal substrates from acids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | (decomposition) |
Physical Form | Crystalline |
Quantity | 50 g |
UN Number | UN3181 |
Solubility Information | Soluble in hot isopropanol,warm hexane and toluene; Tends to precipitate over time. |
Formula Weight | 387.67 |
Sensitivity | Moisture sensitive |
Chemical Name or Material | Zirconium (IV) isopropoxide isopropanol complex |
RUO – Research Use Only