missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Zirconium(IV) hexafluoro-2,4-pentanedionate

Brand:  Alfa Aesar™ 039542.06

Code : NEW

Additional Details : CAS Number : 19530-02-0 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 11368338

  • £85.60 / 5g


Estimated Shipment date
from Supplier 06-02-2018
Add to basket

Specifications

Chemical Identifiers

Zirconium(IV) hexafluoro-2,4-pentanedionate
919.47
Zirconium hexafluoroacetonylacetonate
19530-02-0
MFCD00079660

Specifications

Zirconium(IV) hexafluoro-2,4-pentanedionate
Crystalline
MFCD00079660
Odorless
Off-white
5g
19530-02-0
919.47
Zirconium hexafluoroacetonylacetonate
225°C
40° to 43°C
Videos
SDS