missing translation for 'onlineSavingsMsg'
Learn More

Zirconium(IV) fluoride, low hafnium, 99%, Hf <0.1%, Thermo Scientific Chemicals

Catalog Number p-7048812
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
7783-64-4
MFCD00011307
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
7783-64-4
MFCD00011307

Zirconium fluoride is used as a zirconium source in oxygen-sensitive applications such as metal production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 600°C (sublimation)
Color White
Physical Form Powder
Quantity 25 g
Linear Formula ZrF4
UN Number UN3260
Merck Index 14,10175
Solubility Information Insoluble in water.
Formula Weight 167.21
Percent Purity 99%
Concentration or Composition (by Analyte or Components) Hf <0.1%
Refractive Index 1.59
Sensitivity Moisture sensitive
Density 4.43 g/mL
Chemical Name or Material Zirconium(IV) fluoride, Low hafnium
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'