missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Zirconium Crucible

Brand:  Alfa Aesar™ 035924.KT

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15413595

  • £163.13 / Each


Estimated Shipment date
from Supplier 03-07-2018
Add to basket

Specifications

Specifications

With SlideOn Ring; Ring/ Rim OD: 48mm
42mm
Zirconium Crucible
29mm
44mm
Zirconium
Videos
SDS