missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Zirconium Crucible

Brand:  Alfa Aesar™ 035902.KT

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15443585

  • £164.10 / Each


Estimated Shipment date
from Supplier 07-06-2018
Add to basket

Specifications

Specifications

With Flanged Rim; Ring/ Rim OD: 54mm
47mm
Zirconium Crucible
36mm
35mm
Zirconium
Videos
SDS