missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Zirconium Cover for Crucible

Brand:  Alfa Aesar™ 036005.KT

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15481397

  • £22.10 / Each


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Specifications

5mL
Depth:18mm
Zirconium
For 35888 crucible
Zirconium Crucible Cover
Videos
SDS