missing translation for 'onlineSavingsMsg'
Learn More

Ziprasidone hydrochloride, Tocris Bioscience™

5-HT2A/D2 antagonist; atypical antipsychotic

Brand:  Tocris (Bio-Techne) 308510

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13432364

  • £126.42 / 10mg

Estimated Shipment date
from Supplier 31-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Atypical antipsychotic that displays combined serotonin and dopamine receptor antagonism. Displays high affinity at 5-HT2A receptors with a 5-HT2A/D2 affinity ratio greater than any other clinically available atypical antipsychotics (pKi values are 9.38, 8.88, 8.69, 8.47, 8.32, 8.14, 7.98, 7.49, 7.33 and 6.28 for 5-HT2A, 5-HT2C, 5-HT1D, 5-HT1A, D2, D3, α1, D4, H1 and D1 receptors respectively).
Specifications

Chemical Identifiers

122883-93-6
467.409
ziprasidone hydrochloride monohydrate, ziprasidone hydrochloride, geodon, 5-2-4-benzo d isothiazol-3-yl piperazin-1-yl ethyl-6-chloroindolin-2-one hydrochloride hydrate, unii-216x081oru, ziprasidone hcl hydrate, ziprasidone hydrochloride usan:usp, zeldox, ziprasidone hydrochloride usan
CHEBI:32314
C1CN(CCN1CCC2=C(C=C3C(=C2)CC(=O)N3)Cl)C4=NSC5=CC=CC=C54.O.Cl
C21H24Cl2N4O2S
ZCBZSCBNOOIHFP-UHFFFAOYSA-N
60853
5-[2-[4-(1,2-benzothiazol-3-yl)piperazin-1-yl]ethyl]-6-chloro-1,3-dihydroindol-2-one;hydrate;hydrochloride

Specifications

10mg
122883-93-6
ziprasidone hydrochloride monohydrate, ziprasidone hydrochloride, geodon, 5-2-4-benzo d isothiazol-3-yl piperazin-1-yl ethyl-6-chloroindolin-2-one hydrochloride hydrate, unii-216x081oru, ziprasidone hcl hydrate, ziprasidone hydrochloride usan:usp, zeldox, ziprasidone hydrochloride usan
C1CN(CCN1CCC2=C(C=C3C(=C2)CC(=O)N3)Cl)C4=NSC5=CC=CC=C54.O.Cl
467.409
CHEBI:32314
>99%
Ziprasidone hydrochloride
C21H24Cl2N4O2S
ZCBZSCBNOOIHFP-UHFFFAOYSA-N
5-[2-[4-(1,2-benzothiazol-3-yl)piperazin-1-yl]ethyl]-6-chloro-1,3-dihydroindol-2-one;hydrate;hydrochloride
60853
449.4
Videos
Documents