missing translation for 'onlineSavingsMsg'
Learn More

Zinc titanium oxide, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7051628
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
12036-69-0
O4TiZn2
242.62
MFCD00054107
ZBFOLPMOGPIUGP-UHFFFAOYSA-N
Zinc titanate
This item is not returnable. View return policy
12036-69-0
O4TiZn2
242.62
MFCD00054107
ZBFOLPMOGPIUGP-UHFFFAOYSA-N
Zinc titanate

Zinc titanium oxide is used as a regenerable catalyst, a pigment and a sorbent of sulfur compounds. It is also used as primary and secondary intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Powder
Quantity 50 g
Solubility Information Insoluble in water.
Formula Weight 242.64
Percent Purity 99.9%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Zinc titanium oxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'