missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc tetrafluoroborate hydrate, Zn 18% min, Thermo Scientific™
Click to view available options
Quantity:
10g
50g
250g
Description
Zinc tetrafluoroborate hydrate is used as a precursor for homometallic, trinuclear heteroscorpionate complexes applied in studies of electronic and magnetic properties. Further, it serves as a mild catalyst for epoxide ring opening with amines.
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Quantity | 10g |
Solubility Information | Soluble in water and alcohol. |
Formula Weight | 238.98 (Anhydrous) |
Physical Form | Powder |
Anhydrous or Hydrate | Hydrate |
Sensitivity | Hygroscopic |
Chemical Name or Material | Zinc Tetrafluoroborate |
RUO – Research Use Only