missing translation for 'onlineSavingsMsg'
Learn More

Zinc tetrafluoroborate hydrate, Zn 18% min, Thermo Scientific Chemicals

Catalog Number 11314939
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
27860-83-9
B2F8Zn
238.99
MFCD00150371
BUMIVIYHDVNOQE-UHFFFAOYSA-N
zinc tetrafluoroborate hydrate, 2bf4.zn.h2o, zinc tetrafluoroborate-water 1/2/1, zinc 2+ hydrate ditetrafluoroborate, zinc 2+ ion hydrate ditetrafluoroborate, borate 1-,tetrafluoro-, zinc 2:1 , hexahydrate 9ci
16212112
[Zn++].F[B-](F)(F)F.F[B-](F)(F)F
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
27860-83-9
B2F8Zn
238.99
MFCD00150371
BUMIVIYHDVNOQE-UHFFFAOYSA-N
zinc tetrafluoroborate hydrate, 2bf4.zn.h2o, zinc tetrafluoroborate-water 1/2/1, zinc 2+ hydrate ditetrafluoroborate, zinc 2+ ion hydrate ditetrafluoroborate, borate 1-,tetrafluoro-, zinc 2:1 , hexahydrate 9ci
16212112
[Zn++].F[B-](F)(F)F.F[B-](F)(F)F

Zinc tetrafluoroborate hydrate is used as a precursor for homometallic, trinuclear heteroscorpionate complexes applied in studies of electronic and magnetic properties. Further, it serves as a mild catalyst for epoxide ring opening with amines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Powder
Quantity 50 g
Solubility Information Soluble in water and alcohol.
IUPAC Name zinc;ditetrafluoroborate;hydrate
Formula Weight 238.98 (anhydrous)
Sensitivity Hygroscopic
Chemical Name or Material Zinc Tetrafluoroborate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'