missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc tetrafluoroborate hydrate, Zn 18% min, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Description
Zinc tetrafluoroborate hydrate is used as a precursor for homometallic, trinuclear heteroscorpionate complexes applied in studies of electronic and magnetic properties. Further, it serves as a mild catalyst for epoxide ring opening with amines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Physical Form | Powder |
Quantity | 250 g |
Solubility Information | Soluble in water and alcohol. |
IUPAC Name | zinc(2+) bis(tetrafluoroboranuide) |
Formula Weight | 238.98 (Anhydrous) |
Sensitivity | Hygroscopic |
Chemical Name or Material | Zinc Tetrafluoroborate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'