missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Sulphate Solution, European Pharmacopoeia Grade, Fisher Chemical™
Click to view available options
Quantity:
1000 mL
Specifications
Quantity | 1000 mL |
Safety and Handling
Product Identifier
- Zinc Sulphate 0.0500N (0.05M). Volumetric Solution (RP 406)
- Warning
- Serious eye damage/eye irritation Category 2
- LONG-TERM AQUATIC HAZARD Chronic 3
- H319-Causes serious eye irritation.
- H412-Harmful to aquatic life with long lasting effects.
- P264-Wash thoroughly after handling.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P501-Dispose of contents/container to .
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'