missing translation for 'onlineSavingsMsg'
Learn More

Zinc sulfide, 99.99%, (trace metal basis), < 10 microns, Thermo Scientific Chemicals

Catalog Number p-221519
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1314-98-3
SZn
97.44
MFCD00011301
DRDVZXDWVBGGMH-UHFFFAOYSA-N
zinc sulfide, sachtolith, sphalerite, zinc blende, albalith, cleartran, zinc monosulfide, pigment white 7, zinc sulfide zns, zinc sulphide
14821
[S--].[Zn++]
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1314-98-3
SZn
97.44
MFCD00011301
DRDVZXDWVBGGMH-UHFFFAOYSA-N
zinc sulfide, sachtolith, sphalerite, zinc blende, albalith, cleartran, zinc monosulfide, pigment white 7, zinc sulfide zns, zinc sulphide
14821
[S--].[Zn++]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY
Color White to Yellow
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.99%
Linear Formula ZnS
Merck Index 15, 10360
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acids
IUPAC Name sulfanylidenezinc
Formula Weight 97.44
Percent Purity 99.99%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Zinc sulfide, 99.99%, <10 microns
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'