missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Zinc sulfide, 99.99% (metals basis)

Brand:  Alfa Aesar™ 013239.A1

Code : NEW

Additional Details : CAS Number : 1314-98-3 Weight : 1.00000kg

Quantity 1kg

 View more versions of this product

Product Code. 11306688

  • £779.40 / 1kg


Estimated Shipment date
from Supplier 01-05-2018
Add to basket

Specifications

Chemical Identifiers

Zinc sulfide
ZnS
MFCD00011301
1314-98-3
97.43
14,10160

Specifications

Zinc sulfide
Lumps
(metals basis)
97.43
14,10160
Odorless
White
1kg
1314-98-3
99.99%
ZnS
MFCD00011301
4.10g/mL
2 to 20mm
1700°C
Videos
SDS