missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc sulfate monohydrate, 99%, Thermo Scientific Chemicals
18.80 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | White |
Physical Form | Powder |
Quantity | 250 g |
Assay Percent Range | 98.5% min. (Complexometry) |
Linear Formula | ZnSO4·H2O |
Merck Index | 15, 10359 |
Solubility Information | Solubility in water: 350g/L (20°C). Other solubilities: practically insoluble in ethanol |
Formula Weight | 179.47 |
Percent Purity | 99% |
Identification | Confirmed (Zinc), Confirmed (Sulfate) |
Show More |
Safety and Handling
Product Identifier
- Zinc sulfate monohydrate
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H302-Harmful if swallowed.
- H318-Causes serious eye damage.
- H410-Very toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P273-Avoid release to the environment.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'