missing translation for 'onlineSavingsMsg'
Learn More

Zinc Sulfate Heptahydrate, Extra Pure, SLR, Fisher Chemical™

Brand:  Fisher Chemical Z/1550/53

Code : 21

Additional Details : CAS Number : 7446-20-0 Weight : 0.50000kg Transport : UN number : 3077 Chem class : 9 Pack group : III


Packaging Plastic powder jar
Quantity 500g

 View more versions of this product

Product Code. 10040370

  • £41.54 / 500g

In stock, ready to ship: 18

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Zinc Sulfate Heptahydrate, Extra Pure, SLR

Specifications

Chemical Identifiers

7446-20-0
287.54
RZLVQBNCHSJZPX-UHFFFAOYSA-L
62640
zinc;sulfate;heptahydrate
H14O11SZn
MFCD00149894
zinc sulfate heptahydrate, zinc sulfate jan, zinc sulfate 1:1 heptahydrate, unii-n57ji2k7wp, zinc vitriol heptahydrate, zinc sulfate heptahydrate 1:1:7, white vitriol heptahydrate, ccris 5563, zinc sulfate znso4 heptahydrate, znso4.7h2o
CHEBI:32312
O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O

Specifications

7446-20-0
MFCD00149894
RZLVQBNCHSJZPX-UHFFFAOYSA-L
zinc;sulfate;heptahydrate
62640
287.53g/mol
Plastic powder jar
100°C
500g
H14O11SZn
zinc sulfate heptahydrate, zinc sulfate jan, zinc sulfate 1:1 heptahydrate, unii-n57ji2k7wp, zinc vitriol heptahydrate, zinc sulfate heptahydrate 1:1:7, white vitriol heptahydrate, ccris 5563, zinc sulfate znso4 heptahydrate, znso4.7h2o
O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O
287.54
CHEBI:32312
Solid
White
4.4 to 6
Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1

Hazard Statement
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P273-Avoid release to the environment.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

Supplemental information
  • MIXTURE LIST-Contains : Zinc sulfate heptahydrate, Zinc sulfate

SDS Documents
Certificates