missing translation for 'onlineSavingsMsg'
Learn More

Zinc Sulfate Heptahydrate, Extra Pure, SLR, Fisher Chemical™

Catalog Number p-7073673
Click to view available options
Quantity:
2 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
This item is not returnable. View return policy
7446-20-0
H14O11SZn
287.54
MFCD00149894
RZLVQBNCHSJZPX-UHFFFAOYSA-L
zinc sulfate heptahydrate, zinc sulfate jan, zinc sulfate 1:1 heptahydrate, unii-n57ji2k7wp, zinc vitriol heptahydrate, zinc sulfate heptahydrate 1:1:7, white vitriol heptahydrate, ccris 5563, zinc sulfate znso4 heptahydrate, znso4.7h2o
62640
CHEBI:32312
zinc;sulfate;heptahydrate
O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O
This item is not returnable. View return policy
7446-20-0
H14O11SZn
287.54
MFCD00149894
RZLVQBNCHSJZPX-UHFFFAOYSA-L
zinc sulfate heptahydrate, zinc sulfate jan, zinc sulfate 1:1 heptahydrate, unii-n57ji2k7wp, zinc vitriol heptahydrate, zinc sulfate heptahydrate 1:1:7, white vitriol heptahydrate, ccris 5563, zinc sulfate znso4 heptahydrate, znso4.7h2o
62640
CHEBI:32312
zinc;sulfate;heptahydrate
O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O

Zinc Sulfate Heptahydrate, Extra Pure, SLR

TRUSTED_SUSTAINABILITY
Melting Point 100°C
Color White
pH 4.4 to 6
Physical Form Solid
Quantity 2 kg
Formula Weight 287.53g/mol
Packaging Plastic powder jar
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Zinc sulfate heptahydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P273-Avoid release to the environment.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'