missing translation for 'onlineSavingsMsg'
Learn More

Zinc sulfate heptahydrate, ACS, 99.0-103.0%, Alfa Aesar™

£18.79 - £118.00

Chemical Identifiers

CAS 7446-20-0
Molecular Formula H14O11SZn
Molecular Weight (g/mol) 287.54
MDL Number MFCD00149894
InChI Key RZLVQBNCHSJZPX-UHFFFAOYSA-L
Synonym zinc sulfate heptahydrate, zinc sulfate jan, zinc sulfate 1:1 heptahydrate, unii-n57ji2k7wp, zinc vitriol heptahydrate, zinc sulfate heptahydrate 1:1:7, white vitriol heptahydrate, ccris 5563, zinc sulfate znso4 heptahydrate, znso4.7h2o
PubChem CID 62640
ChEBI CHEBI:32312
IUPAC Name zinc;sulfate;heptahydrate
SMILES O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11397728
SDS View Documents
Alfa Aesar
033399.22
100g
£18.79
100g
View alternative products
Save Up to 
»
In stock, ready to ship: 9
Please to purchase this item. Need a web account? Register with us today!
11387728
SDS View Documents
Alfa Aesar
033399.36
500g
£36.77
500g
View alternative products
Save Up to 
»
In stock, ready to ship: 7
Please to purchase this item. Need a web account? Register with us today!
11455000
SDS View Documents
Alfa Aesar
033399.A4
2.5kg
£118.00
2.50kg
View alternative products
Save Up to 
»
In stock, ready to ship: 2
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

7446-20-0
287.54
RZLVQBNCHSJZPX-UHFFFAOYSA-L
62640
zinc;sulfate;heptahydrate
H14O11SZn
MFCD00149894
zinc sulfate heptahydrate, zinc sulfate jan, zinc sulfate 1:1 heptahydrate, unii-n57ji2k7wp, zinc vitriol heptahydrate, zinc sulfate heptahydrate 1:1:7, white vitriol heptahydrate, ccris 5563, zinc sulfate znso4 heptahydrate, znso4.7h2o
CHEBI:32312
O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O

Specifications

Zinc sulfate
7446-20-0
H14O11SZn
zinc sulfate heptahydrate, zinc sulfate jan, zinc sulfate 1:1 heptahydrate, unii-n57ji2k7wp, zinc vitriol heptahydrate, zinc sulfate heptahydrate 1:1:7, white vitriol heptahydrate, ccris 5563, zinc sulfate znso4 heptahydrate, znso4.7h2o
RZLVQBNCHSJZPX-UHFFFAOYSA-L
zinc;sulfate;heptahydrate
62640
287.54 (161.43 Anhydrous)
ACS
Hygroscopic
100g
heptahydrate
99.0-103.0%
MFCD00149894
Soluble in water,glycerol.
O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O
287.54
CHEBI:32312
Lump
1.97
100°C
SDS
Product Certifications