missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Sulfate Heptahydrate, ≥99.0%, Reagent Grade, Honeywell™
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Poly bottle
Description
- ≥95% for general laboratory use.
Specifications
Melting Point | ∼100°C |
Color | Colorless |
pH | 4.4 to 5.6 (at 20°C) |
Quantity | 2.5 kg |
Linear Formula | ZnSO4 · 7H2O |
UN Number | UN3077 |
Formula Weight | 287.56g/mol |
Percent Purity | ≥99.0% |
Odor | Odorless |
Packaging | Poly bottle |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'