missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Zinc selenide crystal optic rectangle, 41mm x 23mm x 2mm, polished both sides

Brand:  Alfa Aesar™ 038484.KT

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 15436255

  • £590.90 / Each


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Specifications

Zinc selenide crystal optic rectangle
41mm x 23mm x 2mm, Polished both sides
1 each
Videos
SDS