missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Alfa Aesar Zinc selenide crystal optic rectangle, 38.5mm x 19.5mm x 2mm, polished both sides

Brand:  Thermo Scientific Alfa Aesar 038472.KT

Zinc selenide crystal optic rectangle, 38.5mm x 19.5mm x 2mm, polished both sides

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 15453655

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.