missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Zinc selenide crystal optic rectangle, 38.5mm x 19.5mm x 2mm (drilled), polished both sides

Brand:  Alfa Aesar™ 038478.KT

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 15403665

  • £287.20 / Each


Estimated Shipment date
from Supplier 09-07-2018
Add to basket

Specifications

Specifications

Zinc selenide crystal optic rectangle
38.5mm x 19.5mm x 2mm (drilled), Polished both sides
1 each
Videos
SDS