missing translation for 'onlineSavingsMsg'
Learn More

Zinc selenide, 99.99%, (trace metal basis), Thermo Scientific Chemicals

Catalog Number 10650395
Click to view available options
Quantity:
10 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1315-09-9
SeZn
144.35
MFCD00011296
GTLQJUQHDTWYJC-UHFFFAOYSA-N
zinc selenide znse, znse, zinc selenide, spectro, zinc selenide, powder, 10 mum trace metals basis, zinc selenide crystal optic disc, 13mm x 1mm, polished both sides, zinc selenide crystal optic disc, 13mm x 2mm, polished both sides, zinc selenide crystal optic disc, 25mm x 2mm, polished both sides, zinc selenide optical window, 12.7mm 0.5in dia x 1mm 0.04in thick, zinc selenide optical window, 12.7mm 0.5in dia x 2mm 0.08in thick, zinc selenide optical window, 25.4mm 1.0in dia x 1mm 0.04in thick
zinc(2+) selandiide
[Zn++].[Se--]
This item is not returnable. View return policy
1315-09-9
SeZn
144.35
MFCD00011296
GTLQJUQHDTWYJC-UHFFFAOYSA-N
zinc selenide znse, znse, zinc selenide, spectro, zinc selenide, powder, 10 mum trace metals basis, zinc selenide crystal optic disc, 13mm x 1mm, polished both sides, zinc selenide crystal optic disc, 13mm x 2mm, polished both sides, zinc selenide crystal optic disc, 25mm x 2mm, polished both sides, zinc selenide optical window, 12.7mm 0.5in dia x 1mm 0.04in thick, zinc selenide optical window, 12.7mm 0.5in dia x 2mm 0.08in thick, zinc selenide optical window, 25.4mm 1.0in dia x 1mm 0.04in thick
zinc(2+) selandiide
[Zn++].[Se--]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY
Melting Point >1100°C
Color Yellow
Physical Form Shiny Fine Powder
Quantity 10 g
Assay Percent Range 99.99%
Linear Formula ZnSe
Merck Index 15, 10356
Solubility Information Solubility in water: insoluble in water. Other solubilities: decomposes in diluted nitric acid
Formula Weight 144.34
Percent Purity 99.99%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Zinc selenide, 99.99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'