missing translation for 'onlineSavingsMsg'
Learn More

Zinc rod, 10mm (0.39in) dia, 99.9998% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7049077
Click to view available options
Quantity:
50 mm
250 mm
This item is not returnable. View return policy
7440-66-6
Zn
65.38
MFCD00011291
HCHKCACWOHOZIP-UHFFFAOYSA-N
dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
23994
CHEBI:30185
zinc
[Zn]
This item is not returnable. View return policy
7440-66-6
Zn
65.38
MFCD00011291
HCHKCACWOHOZIP-UHFFFAOYSA-N
dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
23994
CHEBI:30185
zinc
[Zn]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 50 mm
Percent Purity 99.9998%
Odor Odorless
Weight ≈5.60g/cm
Form Rod
Assay (metals basis)
Chemical Name or Material Zinc rod, 10mm (0.39 in.) dia.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'