Learn More
Thermo Scientific Alfa Aesar Zinc rod, 1.27cm (0.5 in.) dia. x 15cm (5.9 in.) long, 99.5% (metals basis)
Brand: Thermo Scientific Alfa Aesar 010434.KK
Zinc rod, 1.27cm (0.5 in.) dia. x 15cm (5.9 in.) long, 99.5% (metals basis)
Code : NEW
Additional Details : CAS Number : 7440-66-6 Weight : 0.01000kg