missing translation for 'onlineSavingsMsg'
Learn More

Zinc protoporphyrin IX, Tocris Bioscience™

Heme oxygenase and guanylyl cyclase inhibitor

Brand:  Tocris (Bio-Techne) 074650

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13457943

  • £282.49 / 50mg


Estimated Shipment
06-03-2019
Add to basket

Description

Description

Inhibitor of heme oxygenase, which generates the putative neurotransmitter CO.
Specifications

Chemical Identifiers

15442-64-5
626.034
zinc protoporphyrin
zinc;3-[18-(2-carboxyethyl)-8,13-bis(ethenyl)-3,7,12,17-tetramethyl-23H-porphyrin-21-id-2-yl]propanoate
C34H32N4O4Zn
FUTVBRXUIKZACV-UHFFFAOYSA-L
131664383
CC1=C(C2=CC3=NC(=CC4=C(C(=C(N4)C=C5C(=C(C(=N5)C=C1[N-]2)C)C=C)C)C=C)C(=C3CCC(=O)O)C)CCC(=O)[O-].[Zn+2]

Specifications

Zinc protoporphyrin IX
C34H32N4O4Zn
FUTVBRXUIKZACV-UHFFFAOYSA-L
zinc;3-[18-(2-carboxyethyl)-8,13-bis(ethenyl)-3,7,12,17-tetramethyl-23H-porphyrin-21-id-2-yl]propanoate
131664383
>98%
15442-64-5
zinc protoporphyrin
CC1=C(C2=CC3=NC(=CC4=C(C(=C(N4)C=C5C(=C(C(=N5)C=C1[N-]2)C)C=C)C)C=C)C(=C3CCC(=O)O)C)CCC(=O)[O-].[Zn+2]
626.034
626.03
50mg
Videos