missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Zinc phosphate (ortho), Puratronic™, 99.995% (metals basis)

Brand:  Alfa Aesar™ 013013.14

Code : NEW

Additional Details : CAS Number : 7779-90-0 Weight : 0.02500kg Transport : UN number : 3077 Chem class : 9 Pack group : III

Quantity 25g

 View more versions of this product

Product Code. 11302118

  • £210.40 / 25g


EU Stock 1
Add to basket

Specifications

Chemical Identifiers

Zinc phosphate (ortho)
Puratronic™
386.05
UN3077
7779-90-0
Zn3(PO4)2
MFCD00036282
Zinc orthophosphate

Specifications

Zinc phosphate (ortho)
Puratronic™
99.995%
Zn3(PO4)2
MFCD00036282
Zinc orthophosphate
Odorless
25g
7779-90-0
Powder
(metals basis)
386.05
UN3077
3.75g/mL
White
Videos
SDS