missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc phosphate hydrate, tech., Thermo Scientific Chemicals
96.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
500 g
2 kg
Description
Zinc phosphate hydrate is used in dentistry. It is commonly used for luting permanent metal restorations and as a base for dental restorations. Zinc phosphate cement is also used for cementation of inlays, crowns, bridges, and orthodontic appliances and occasionally as a temporary restoration.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Physical Form | Powder |
Quantity | 2 kg |
Solubility Information | Soluble in acetic acid,dilute mineral acids,ammonia. Insoluble in water |
SMILES | [O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Zn+2].[Zn+2].[Zn+2] |
IUPAC Name | trizinc;diphosphate |
Formula Weight | 386.05 (Anhydrous) |
Grade | Technical |
Chemical Name or Material | Zinc phosphate hydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'