missing translation for 'onlineSavingsMsg'
Learn More

Zinc perchlorate hexahydrate, Reagent Grade, Thermo Scientific Chemicals

Catalog Number 11317367
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
10025-64-6
Cl2H12O14Zn
372.362
MFCD00150370
PADPILQDYPIHQQ-UHFFFAOYSA-L
zinc perchlorate hexahydrate, perchloric acid, zinc salt, hexahydrate, 2clo4.zn.6h2o, zinc diperchlorate hexahydrate, zinc perchlorate 6h2o, zincperchloratehexahydrate, zinc 2+ hexahydrate diperchlorate, zinc perchlorate hexahydrate, reagent grade, zinc, ion zn2+ hexahydrate diperchlorate ion, zinc perchlorate hexahydrate trace metals basis 10g
202253
zinc;diperchlorate;hexahydrate
O.O.O.O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Zn+2]
This item is not returnable. View return policy
10025-64-6
Cl2H12O14Zn
372.362
MFCD00150370
PADPILQDYPIHQQ-UHFFFAOYSA-L
zinc perchlorate hexahydrate, perchloric acid, zinc salt, hexahydrate, 2clo4.zn.6h2o, zinc diperchlorate hexahydrate, zinc perchlorate 6h2o, zincperchloratehexahydrate, zinc 2+ hexahydrate diperchlorate, zinc perchlorate hexahydrate, reagent grade, zinc, ion zn2+ hexahydrate diperchlorate ion, zinc perchlorate hexahydrate trace metals basis 10g
202253
zinc;diperchlorate;hexahydrate
O.O.O.O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Zn+2]

Zinc perchlorate hexahydrate, efficiently catalyses the esterification between nearly equimolar amounts of carboxylic acids and alcoholsZinc perchlorate hexahydrate [Zn (ClO4) 2· 6H2O] was found to be a highly efficient catalyst for acylation of electron deficient phenols, sterically hindered alcohols and electron deficient and sterically hindered amines. Zinc (II) perchlorate hexahydrate has been found to be a new and efficient catalyst for conjugate addition of thiols to α, β-unsaturated ketones under solvent-free conditions at room temperature.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 106°C
Color White
Physical Form Crystalline
Quantity 100 g
Linear Formula Zn(ClO4)2·6H2O
UN Number UN1481
Merck Index 14,10148
Solubility Information Soluble in water, alcohol
Formula Weight 372.37 (264.28 Anhydrous)
Grade Reagent
Odor Odorless
Refractive Index 1.5
Sensitivity Hygroscopic
Density 2.25 g/mL
Chemical Name or Material Zinc perchlorate hexahydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'