Learn More
Zinc perchlorate hexahydrate, Reagent Grade, Thermo Scientific Chemicals
Description
Zinc perchlorate hexahydrate, efficiently catalyses the esterification between nearly equimolar amounts of carboxylic acids and alcoholsZinc perchlorate hexahydrate [Zn (ClO4) 2· 6H2O] was found to be a highly efficient catalyst for acylation of electron deficient phenols, sterically hindered alcohols and electron deficient and sterically hindered amines. Zinc (II) perchlorate hexahydrate has been found to be a new and efficient catalyst for conjugate addition of thiols to α, β-unsaturated ketones under solvent-free conditions at room temperature.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 106°C |
Color | White |
Physical Form | Crystalline |
Quantity | 500 g |
Linear Formula | Zn(ClO4)2·6H2O |
UN Number | UN1481 |
Merck Index | 14,10148 |
Solubility Information | Soluble in water, alcohol |
Formula Weight | 372.37 (264.28 Anhydrous) |
Grade | Reagent |
Show More |
RUO – Research Use Only