missing translation for 'onlineSavingsMsg'
Learn More

Zinc Oxide USP Powder, Macron Fine Chemicals™

macron

Brand:  Macron Fine Chemicals 8824-05

Additional Details : Weight : 2.50000kg

Packaging STAKMOR
Quantity 2.5kg

 View more versions of this product

Product Code. 15598484

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

1314-13-2
81.379
zinc oxide, zinc white, amalox, permanent white, chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
CHEBI:36560
O=[Zn]
OZn
XLOMVQKBTHCTTD-UHFFFAOYSA-N
14806
oxozinc

Specifications

Zinc Oxide
OZn
XLOMVQKBTHCTTD-UHFFFAOYSA-N
oxozinc
14806
81.39
USP
2.5kg
1314-13-2
zinc oxide, zinc white, amalox, permanent white, chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
O=[Zn]
81.379
CHEBI:36560
Powder
STAKMOR
Videos
Documents