missing translation for 'onlineSavingsMsg'
Learn More

Zinc Oxide, puriss. p.a., ACS Reagent, ≥99% (KT), Honeywell Fluka™

Catalog Number 15637580
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
1314-13-2
OZn
81.38
MFCD00011300
XLOMVQKBTHCTTD-UHFFFAOYSA-N
zinc oxide, zinc white, amalox, permanent white, Chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
14806
CHEBI:36560
oxozinc
O=[Zn]
This item is not returnable. View return policy
1314-13-2
OZn
81.38
MFCD00011300
XLOMVQKBTHCTTD-UHFFFAOYSA-N
zinc oxide, zinc white, amalox, permanent white, Chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
14806
CHEBI:36560
oxozinc
O=[Zn]

Puriss. p.a., ACS Reagent,≥99.0% (KT)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Melting Point ∼1975°C
Color White
pH ∼7 (at 20°C)
Quantity 1 kg
Linear Formula ZnO
UN Number UN3077
Formula Weight 81.39g/mol
Percent Purity ≥99% (KT)
Packaging Poly Bottle
Appearance Powder
Density 5.6g/cm3 at 20°C
Chemical Name or Material Zinc oxide
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'