missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Zinc oxide, nanopowder, 99+% (metals basis)

Brand:  Alfa Aesar™ 044299.09

Code : NEW

Additional Details : CAS Number : 1314-13-2 Weight : 0.01000kg Transport : UN number : 3077 Chem class : 1 Pack group : III

Quantity 10g

 View more versions of this product

Product Code. 11468390

  • £230.30 / 10g


Estimated Shipment date
from Supplier 27-02-2018
Add to basket

Specifications

Chemical Identifiers

Zinc oxide
ZnO
MFCD00011300
14,10147
1314-13-2
81.37
UN3077

Specifications

Zinc oxide
Nanopowder
(metals basis)
81.37
UN3077
5.606g/mL
7 to 13nm APS
10g
1314-13-2
≥99%
ZnO
MFCD00011300
14,10147
Odorless
1975°C
Videos
SDS