missing translation for 'onlineSavingsMsg'
Learn More

Zinc oxide, NanoArc™ ZN-0605, Thermo Scientific Chemicals

Catalog Number p-7059980
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
1314-13-2
OZn
81.38
MFCD00011300
XLOMVQKBTHCTTD-UHFFFAOYSA-N
zinc oxide, zinc white, amalox, permanent white, Chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
14806
CHEBI:36560
oxozinc
O=[Zn]
This item is not returnable. View return policy
1314-13-2
OZn
81.38
MFCD00011300
XLOMVQKBTHCTTD-UHFFFAOYSA-N
zinc oxide, zinc white, amalox, permanent white, Chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
14806
CHEBI:36560
oxozinc
O=[Zn]

Employed in the preparation of NaZnSiO3OH, a novel chiral framework material which has potential application in ion exchange, adsorption or catalysis. Used in electronic industry, instrumental industry, manufacture of electrical device, image recorder, rheostat, phosphor. Used as a sun screening agent in cosmetics, antibacterial and health protection antiager; UV protection; Piezoelectrics; High-temperature lubricant in gas turbine engines; Flame retardant; Adsorption; Dental cements; Environmental remediation; Gas sensors; Photocatalytic decontamination; Attenuation of ultraviolet light; Demilitarization of chemical and biological warfare agents; Cosmetics and cosmeceuticals; Electrodes for solar cells; Varistors; Pigments for paints.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 1975°C
Color White
Physical Form Powder
Quantity 100 g
Linear Formula ZnO
UN Number UN3077
Merck Index 14,10147
Solubility Information Insoluble in water.
Formula Weight 81.37
Odor Odorless
Chemical Name or Material Zinc oxide NanoArc™ ZN-2605
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'