Learn More
Zinc oxide based sulfur removal material, HiFUEL|r A310, Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 045469.A1
110.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : NEW
Additional Details : Weight : 1.00000kg Transport : UN number : 3077 Chem class : 9 Pack group : III
Description
Zinc oxide is used in laser diodes and light emitting diodes. It helps in removal of sulfur and sulfur compounds from a variety of fluids and gases particularly industrial flue gases.It acts as a precursor to other zinc compounds and additive to protect the rubber from fungi and UV light. It is mixed with stearic acid and used in the vulcanization of rubber. It is used in products like baby powder, barrier creams to treat diaper rashes, calamine cream, anti-dandruff shampoos, and antiseptic ointments. It is used in nuclear pressurized water reactors to prevent corrosion. It acts as a food additive in cereals and prepackaged foods.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Zinc oxide based sulfur removal material HiFUEL™ A310 | |
1 kg | |
Soluble in hydrochloric acid. Insoluble in water. |
MFCD00011300 | |
EM>6, | |
Spheres |
RUO – Research Use Only