missing translation for 'onlineSavingsMsg'
Learn More

Zinc oxide, ACS reagent, Thermo Scientific Chemicals

Catalog Number p-3777116
Click to view available options
Quantity:
5 g
100 g
500 g
1 kg
5 kg
Packaging:
Glass Bottle
Plastic bottle
Plastic drum
This item is not returnable. View return policy
1314-13-2
OZn
81.38
MFCD00011300
XLOMVQKBTHCTTD-UHFFFAOYSA-N
zinc oxide, zinc white, amalox, permanent white, Chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
14806
CHEBI:36560
oxozinc
O=[Zn]
This item is not returnable. View return policy
1314-13-2
OZn
81.38
MFCD00011300
XLOMVQKBTHCTTD-UHFFFAOYSA-N
zinc oxide, zinc white, amalox, permanent white, Chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
14806
CHEBI:36560
oxozinc
O=[Zn]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 1975.0°C
Color White
Physical Form Powder
Quantity 5 g
Linear Formula ZnO
Fieser 01,1294; 05,622
Merck Index 15,10347
Solubility Information Solubility in water: 1.6 mg/l (29°C). Other solubilities: soluble in acids and alkalies, soluble in diluted acetic or mineral acids, ammoni, ammonium carbonate, fixed alkali hydroxide solutio, insoluble in alcohol
Formula Weight 81.38
Percent Purity ≥99.0%
Grade ACS Reagent
Packaging Glass Bottle
Specific Gravity 5.6
Density 5.6000g/mL
Chemical Name or Material Zinc oxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'