missing translation for 'onlineSavingsMsg'
Learn More

Zinc oxalate dihydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7049130
74.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
55906-21-3
MFCD00150611
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
55906-21-3
MFCD00150611

Zinc oxalate dihydrate is used as an chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Physical Form Powder
Quantity 25 g
Molecular Formula C2H6O6Zn
Linear Formula ZnC2O4·2H2O
UN Number UN3288
Merck Index 14,10146
Synonym zinc oxalate dihydrate, zinc oxalate, c2o4zn.2h2o
Solubility Information Soluble in dilute mineral acids.
InChI Key PLYRAVNBPDIEOH-UHFFFAOYSA-N
SMILES C(=O)(C(=O)O)O.O.O.[Zn]
IUPAC Name oxalic acid;zinc;dihydrate
Molecular Weight (g/mol) 191.44
PubChem CID 91886451
Formula Weight 189.42 (153.39 Anhydrous)
Percent Purity 99.999%
Grade Puratronic™
Density 3.28 g/mL
Assay (metals basis)
Chemical Name or Material Zinc oxalate dihydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'