missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Nitrate Hexahydrate, Technical, Fisher Chemical™
Click to view available options
Quantity:
250 g
500 g
Specifications
Melting Point | 36°C |
Color | White |
pH | 5.1 |
Physical Form | Solid |
Quantity | 500 g |
IUPAC Name | zinc(2+) hexahydrate dinitrate |
Formula Weight | 297.46g/mol |
Packaging | Plastic powder jar |
Safety and Handling
Product Identifier
- Zinc nitrate hexahydrate
- Danger
- Oxidising solid Category 2
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 2
- H272-May intensify fire; oxidiser.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H302-Harmful if swallowed.
- H411-Toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P273-Avoid release to the environment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'